IL9
Catalog # BIRBORB97800-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 0.2 ng/ml, corresponding to a specific activity5x106 IU/mg, determined by the dose dependent stimulation of human MO7e cells.
- UniProtKB:P15248
- Protein/peptide name:IL9
- Purity:985% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized IL-9 inbidest water not less than 100 g/ml, which can then befurther diluted to other aqueous solutions. For long termstorage it is recommended to add a carrier protein (0.1%HSA or BSA).
- Molecular weight:14 kDa
- Formulation:Lyophilized