Mouse recombinant IL6 (from E. coli)
Catalog # BIRBORB97798-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 0.02 ng/ml, corresponding to a specific activity5x107 IU/mg, determined by the dose dependent stimulation of murine 7TD1 cells.
- Protein synonyms:Interleukin BSF 2
- UniProtKB:P08505
- Protein/peptide name:IL6
- Purity:90% pure by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Interleukin 6 in 20 mM HCl to 0.1-1.0 mg/ml to regain fullactivity and can then be further diluted to other aqueoussolutions.For long term storage it is recommended to add a carrierprotein (0.1% HSA or BSA
- Molecular weight:22 kDa
- Formulation:Lyophilized. IL-6 is lyophilized from a 1 mg/ml solutioncontaining 10 mM NaAcetate pH 6.0.