Recombinant OPG
BIRBORB97786-50
: Biorbyt
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at−20 °C.
- Protein synonyms:TNFRSF11B
- UniProtKB:O00300
- Protein/peptide name:OPG
- Purity:80% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized Osteoprotegerin in sterile 18MΩ-cm water not lessthan 100 g/ml, which can then be further dilutedto other aqueous solutions.For long term storage it isrecommended to add a carrier protein (0.1% HSA or
- Molecular weight:31 kDa
- Formulation:Lyophilized. The protein was lyophilized from a concentrated (1mg/ml) solution with 1X PBS, 0.1% SDS and1mM DTT.