Order Entry
Hungary
ContactUsLinkComponent
HCV-NS3-S1c
HCV-NS3-S1c
Catalog # BIRBORB97912-100
Supplier:  Biorbyt
CAS Number:  
HCV-NS3-S1c
Catalog # BIRBORB97912-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HCV-NS3-S1c
  • Purity:
    Greater than 95% by SDS-PAGE
  • Formulation:
    Liquid. Supplied in 1.5M urea, 25mM Tris-HCl pH 8,0.2% Triton-X and 50% glycerol.
  • Tested applications:
    ELISA , WB

Specifications

About this item