Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant TrkA Fc
Human recombinant TrkA Fc
Human recombinant TrkA Fc
Catalog # BIRBORB83296-100
Supplier:
Biorbyt
CAS Number:
Expand image
Human recombinant TrkA Fc
Catalog # BIRBORB83296-100
Supplier:
Biorbyt
CAS Number:
Specifications
Pk:
100 µG
Protein/peptide type:
Recombinant
Species:
Human
Protein/peptide name:
TrkA Fc
Tested applications:
WB , SDS-PAGE
Specifications
About this item
Product Details
Documentation
Product Details
Documentation