Order Entry
Hungary
ContactUsLinkComponent
Mouse recombinant IL6 (from E. coli)
Mouse recombinant IL6 (from E. coli)
Catalog # BIRBORB82044-10
Supplier:  Biorbyt
CAS Number:  
Mouse recombinant IL6 (from E. coli)
Catalog # BIRBORB82044-10
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Protein synonyms:
    Interleukin BSF 2
  • Protein/peptide name:
    IL6
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.

Specifications

About this item