Human recombinant TPO (from baculovirus (insect cells))
Catalog # BIRBORB81916-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Protein synonyms:MSA
- Protein/peptide name:TPO
- Formulation:Lyophilised
- Tested applications:ELISA , WB
- Purification:Affinity purified