Human recombinant Epidermal Growth Factor (from E. coli)
Catalog # BIRBORB82036-0.5
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:0,5 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:EGF ELISA Kit
- Protein/peptide name:Epidermal Growth Factor
- Purity:Purified.
- Formulation:Lyophilised.