Order Entry
Hungary
ContactUsLinkComponent
Human alpha 2 antiplasmin (from plasma)
Human alpha 2 antiplasmin (from plasma)
Catalog # BIRBORB82118-100
Supplier:  Biorbyt
CAS Number:  
Human alpha 2 antiplasmin (from plasma)
Catalog # BIRBORB82118-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
    Plasma
  • Species:
    Human
  • Protein synonyms:
    Alpha2-antiplasmin
  • Protein/peptide name:
    Alpha 2 Antiplasmin
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.
  • Tested applications:
    SDS-PAGE

Specifications

About this item