Order Entry
Hungary
ContactUsLinkComponent
Complement C1q (from plasma)
Complement C1q (from plasma)
Catalog # BIRBORB82272-1
Supplier:  Biorbyt
CAS Number:  
Complement C1q (from plasma)
Catalog # BIRBORB82272-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
    Plasma
  • Protein/peptide name:
    Complement C1q
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.

Specifications

About this item