Order Entry
Hungary
ContactUsLinkComponent
 
Toxoplasma gondii recombinant Toxoplasma gondii p30 (from Escherichia coli)
  BIRBORB82475-1
 :  Biorbyt
 :  
Toxoplasma gondii recombinant Toxoplasma gondii p30 (from Escherichia coli)
  BIRBORB82475-1
 :  Biorbyt
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Toxoplasma gondii
  • Protein synonyms:
    SAG1
  • Protein/peptide name:
    Toxoplasma gondii p30
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.
  • Tested applications:
    ELISA , WB