Order Entry
Hungary
ContactUsLinkComponent
Recombinant Plasmodium vivax MSP-1 protein (malaria MSP-1, pv) (from E. coli)
Recombinant Plasmodium vivax MSP-1 protein (malaria MSP-1, pv) (from E. coli)
Catalog # BIRBORB82358-1
Supplier:  Biorbyt
CAS Number:  
Recombinant Plasmodium vivax MSP-1 protein (malaria MSP-1, pv) (from E. coli)
Catalog # BIRBORB82358-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Protein synonyms:
    Plasmodium vivax MSP-1
  • Protein/peptide name:
    Plasmodium vivax MSP-1 protein (Malaria MSP-1, pv)
  • Purity:
    Purified.
  • Formulation:
    Preservative: 0.1% Sodium Azide
  • Tested applications:
    ELISA

Specifications

About this item