Order Entry
Hungary
ContactUsLinkComponent
 
Recombinant Plasmodium vivax CSP protein (malaria CSP pv) (from E. coli)
Catalog # BIRBORB82359-1
Supplier:  Biorbyt
CAS Number:  
Recombinant Plasmodium vivax CSP protein (malaria CSP pv) (from E. coli)
Catalog # BIRBORB82359-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Protein synonyms:
    Plasmodium vivax CSP
  • Protein/peptide name:
    Plasmodium vivax CSP protein (malaria CSP pv)
  • Purity:
    Purified.
  • Formulation:
    Preservative: 0.1% Sodium Azide
  • Tested applications:
    ELISA , LF

Specifications

About this item