Order Entry
Hungary
ContactUsLinkComponent
P. falciparum recombinant Plasmodium falciparum (malaria) histidine-rich protein 2 (HRP-2) (from E. coli)
P. falciparum recombinant Plasmodium falciparum (malaria) histidine-rich protein 2 (HRP-2) (from E. coli)
Catalog # BIRBORB82395-1
Supplier:  Biorbyt
CAS Number:  
P. falciparum recombinant Plasmodium falciparum (malaria) histidine-rich protein 2 (HRP-2) (from E. coli)
Catalog # BIRBORB82395-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    P. falciparum
  • Protein synonyms:
    Plasmodium falciparum (malaria) Histidine-Rich 2 (HRP-2)
  • Protein/peptide name:
    Plasmodium falciparum (malaria) Histidine-Rich Protein 2 (HRP-2)
  • Purity:
    Purified.
  • Formulation:
    Preservative: 0.1% Sodium Azide
  • Tested applications:
    ELISA

Specifications

About this item