Recombinant west nile virus pre-M protein, N-terminal (from E. coli)
BIRBORB82577-100
: Biorbyt
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein synonyms:West Nile Virus Pre-M
- Protein/peptide name:West Nile Virus Pre-M Protein, N-terminal
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB , LF