Human SDF1 alpha (fromE. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The specific activity as determined by its ability to chemoattract human peripheral cells activated with PHA and IL-2 using concentation of 20-80 ng/ml.
- Protein synonyms:C-X-C motif chemokine 12
- Protein/peptide name:SDF1 alpha
- Purity:> 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Stromal Cell-Derived Factor-1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Lys-Pro-Val-Ser-Leu.
- Formulation:The protein was lyophilized from concentrated (1 mg/ml) sterile solution containing no additives.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together