Human recombinant ubiquitin biotin (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized RPS27A although stable at 15 °C for week, should be stored desiccated below −18 °C. For long term storage, to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Ubiquitin
- Protein/peptide name:Ubiquitin Biotin
- Purity:RPS27A Protein biotinilation is determined by Western Blotting and ELISA analysis using streptavidinCHRP conjugated as detection reagent. Free biotin is eliminated by dialysis against PBS. Protein concentration is determined by 280nm absorbance.
- Formulation:The RPS27A is supplied in 1x PBS and 0.05% PBS.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together