Order Entry
Hungary
ContactUsLinkComponent
Human recombinant SUMO2 GST (from E. coli)
Human recombinant SUMO2 GST (from E. coli)
Gyártó:  Biorbyt
Tanúsítványok
Human recombinant SUMO2 GST (from E. coli)
Gyártó:  Biorbyt

Specifikációk

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Protein synonyms:
    Small ubiquitin-related modifier 2
  • Protein/peptide name:
    SUMO2 GST
  • Formulation:
    SUMO2 at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifikációk

Kapcsolódó információk

Frequently Bought Together