Order Entry
Hungary
ContactUsLinkComponent
Human recombinant S100A1 (from E. coli)
Human recombinant S100A1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant S100A1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Biological activity:
    ~3 nmol/mg/min using phosphatidylinositol as substrate.
  • Protein synonyms:
    Protein S100-A1
  • Protein/peptide name:
    S100A1
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Formulation:
    S100A1 in 20 mM Tris-HCl, pH7.0 and 0.1mM EDTA.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together