Recombinant E.coli TRX (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Storage conditions:TRX although stable at 4 °C for weeks, should be stored desiccated below −18 °C. Please prevent freeze thaw cycles.
- Biological activity:TRX activity is assayed by measuring the change in absorbance at 650 nm at 25C using 0.13uM bovine insulin containing 0.33mM DTT (pH 6.5).The specific activity was found to be 3IU/mg.
- Protein synonyms:ADF
- Protein/peptide name:E.coli TRX
- Purity:Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized TRX in sterile H2O.
- Sequence:HMSDKIIHL TDDSFDTDVLKADGAIL VDFW AEWCGPCKMIAPILDEI GKLTVAKLNIDQNPGTAPKYGIRGIPTLLLFKNGEVAATKVGAL DANLA.
- Formulation:Each mg of protein contains 20 mM phosphate buffer pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together