Order Entry
Hungary
ContactUsLinkComponent
 
Human recombinant MAGEA1 (from E. coli)
 :  Biorbyt
 
Human recombinant MAGEA1 (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Protein synonyms:
    Melanoma-associated antigen 1
  • Protein/peptide name:
    MAGEA1
  • Formulation:
    MAGE-1 in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together