Human recombinant PGP9.5 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:Epididymis luminal 117
- Protein/peptide name:PGP9.5
- Formulation:PGP9.5 at 0.1mg/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA, 20% Glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together