Human recombinant KRT8 GST (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:Keratin type II cytoskeletal 8
- Protein/peptide name:KRT8 GST
- Formulation:Cytokeratin-8 in 50mM Tris-Acetate pH-7.5, 1mM EDTA and 20% Glyecrol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together