Human VEGF receptor 1 D1-4 (from Insect cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Insect cells
- Species:Human
- Storage conditions:Lyophilized FLT-1 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution FLT1 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:The activity of FLT1D1-4 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces, and in Far-Western and cross-linking experiments with iodinated VEGF.
- Protein synonyms:Vascular endothelial growth factor A
- Protein/peptide name:VEGF receptor 1 D1-4
- Purity:Greater than 90.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized FLT1 D4 in sterile water not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:FLT1 D1-4 was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together