Recombinant HSV-1 gD
: Biorbyt
- Protein/peptide type:Recombinant
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein synonyms:HSV1
- Protein/peptide name:HSV-1 gD
- Purity:HSV-1 gD protein is >95% pure as determined by 10% PAGE (coomassie staining). HSV-1 gD was purified by proprietary chromatographic technique.
- Formulation:25mM Tris-HCl, 1mM EDTA, and 50% glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together