Mouse recombinant PDGF AA
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected ED50 for this effect is 8-10 ng/ml.
- Protein synonyms:Platelet-Derived Growth Factor-AA
- UniProtKB:P20033
- Protein/peptide name:PDGF AA
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized PDGF-AA in sterile H2O at concentration ranging between 0.1-0.5mg per 1ml, which can then be further diluted to other aqueous solutions.
- Sequence:MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATSNLN PDHREEETGR RRESGKNRKR KRLKPT
- Formulation:The protein was lyophilized with no additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together