Order Entry
Hungary
ContactUsLinkComponent
 
Human recombinant vaspin
 :  Biorbyt
 
Human recombinant vaspin
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Data Not Available.
  • Gene ID:
    145264
  • Protein synonyms:
    Visceral adipose tissue-derived serpin
  • UniProtKB:
    Q8IW75
  • Protein/peptide name:
    Vaspin
  • Purity:
    >98%
  • Sequence:
    MLKPSFSPRN YKALSEVQGW KQRMAAKELA RQNMDLGFKL LKKLAFYNPG RNIFLSPLSI STAFSMLCLG AQDSTLDEIK QGFNFRKMPE KDLHEGFHYI IHELTQKTQD LKLSIGNTLF IDQRLQPQRK FLEDAKNFYS AETILTNFQN LEMAQKQIND FISQKTHGKI NNLIENIDPG TVMLLANYIF FRARWKHEFD PNVTKEEDFF LEKNSSVKVP MMFRSGIYQV GYDDKLSCTI LEIPYQKNIT AIFILPDEGK LKHLEKGLQV DTFSRWKTLL SRRVVDVSVP RLHMTGTFDL KKTLSYIGVS KIFEEHGDLT KIAPHRSLKV GEAVHKAELK MDERGTEGAA GTGAQTLPME TPLVVKIDKP YLLLIYSEKI PSVLFLGKIV NPIGK
  • Formulation:
    Vaspin in 20 mM Tris pH-8, 0.2 um PMSF and 10% glycerol.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together