Human recombinant LFA 3 (from ovarian cell)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Ovarian cell
- Species:Human
- Storage conditions:Lyophilized LFA3 Human although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Human LFA-3 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:CD58
- Protein/peptide name:LFA 3
- Purity:Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized LFA-3 Human in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:Each mg of CD58 contains 0.8mg sucrose, 0.3mg glycine, 0.2 umg sodium citrate dihydrate, and 4ug citric acid monohydrate.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together