Order Entry
Hungary
ContactUsLinkComponent
Rat recombinant IL5
Rat recombinant IL5
Supplier:  Biorbyt
Certificates
Rat recombinant IL5
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is < 0.5 ng/ml, corresponding to a specific activity of > 2 x 106 units/mg.
  • Protein synonyms:
    EDF
  • UniProtKB:
    Q08125
  • Protein/peptide name:
    IL5
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleikin-5 Rat in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MEIPMSTVVK ETLIQLSTHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEILFQNL SLIKKYIDGQ KEKCGEERRK TRHFLDYLQE FLGVMSTEWA MEV
  • Formulation:
    Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together