Order Entry
Hungary
ContactUsLinkComponent
Rat Recombinant MIP1A
Rat Recombinant MIP1A
Supplier:  Biorbyt
Certificates
Rat Recombinant MIP1A
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Assay#1:  Determined by its ability to chemoattract rat peritoneal macrophages using a concentration of 50.0-100.0 ng/ml. Assay#2:  Determined by its ability to chemoattract human blood monocytes using a concentration range of 10.0-100.0 ng/ml.
  • Protein synonyms:
    Macrophage Inflammatory-1 alpha
  • UniProtKB:
    P50229
  • Protein/peptide name:
    MIP1A
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Macrophage Inflammatory Protein-1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    APYGADTPTA CCFSYGRQIP RKFIADYFET SSLCSQPGVI FLTKRNRQIC ADPKETWVQE YITELELNA
  • Formulation:
    The protein was lyophilized from mg/ml solution containing no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together