Bradykinin ELISA kit
Catalog # ENZOADI900206
Gyártó: ENZO LIFE SCIENCES
CAS szám:
Specifikációk
- Pk:1 KIT
- Assay Type:Competitive
- Description:Bradykinin ELISA kit, 96 wells
- Seq:02
- Sensitivity:24,8 pg/ml (range 11,7 - 30000 pg/ml)
Specifikációk
Termék információk
The Bradykinin ELISA kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Bradykinin in plasma, serum and urine samples.
- Sensitive - measure as little as 24,8 pg/ml of Bradykinin
- Rapid results in just 3 hours from up to 41 samples in duplicate
- Broad dynamic range to easily quantify high and low levels of Bradykinin
- Easy to use liquid colour coded reagents reduce errors
For the quantitative determination of species independent Bradykinin in plasma, serum and urine samples.
The Bradykinin EIA kit is a colorimetric, competitive immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP.
Delivery: The kit contains microtiter plate, conjugate, antibody, assay buffer 16, wash buffer concentrate, standard, TMB substrate, stop solution 2, and Streptavidin-HRP.