Order Entry
Hungary
ContactUsLinkComponent
IL-12p70 (human) ELISA kit
  ENZOADI900202
 :  
IL-12p70 (human) ELISA kit
  ENZOADI900202
 :  
  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-12 (p70)
  • Description:
    IL-12p70 ELISA kit (human), 96 wells
  • Seq:
    02
The IL-12p70 (human) ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human IL-12p70 in culture supernatants, plasma, and serum samples.
  • Specific - no cross reactivity to p40 subunit
  • Rapid - results within 3 hours
  • Sensitive - reliably measure as little as 0,9 pg/ml of IL-12p70
  • Non radioactive - enzymatic assay format is safe and reliable
  • Easy to use liquid colour coded reagents reduce errors
  • Ready to use reagents and pre-coated plates reduce errors
 : Kit contains microtiter plate, conjugate, antibody, assay buffer 28, wash buffer concentrate, standard, TMB substrate, and stop solution 2.