Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cocoa butter
Cocoa butter, waxy solid
Cocoa butter, waxy solid
ICNA0290541790
:
MP Biomedicals
:
Cocoa butter, waxy solid
ICNA0290541790
:
MP Biomedicals
:
Pk:
500 g
Seq:
3645
Identity Test:
Passes
:
:
32.1 °C (lit.)
:
Ambient
:
8002-31-1
:
MFCD00146492