Order Entry
Hungary
ContactUsLinkComponent
Human prolactin ELISA kit
Human prolactin ELISA kit
Catalog # BIRBORB138057-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human prolactin ELISA kit
Catalog # BIRBORB138057-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human Prolactin ELISA Kit
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human Prolactin ELISA Kit for the quantitative detection of human Prolactin in cell culture supernates, serum and plasma (heparin, citrate).