Order Entry
Hungary
ContactUsLinkComponent
Human TNFSF12 ELISA kit
Human TNFSF12 ELISA kit
Catalog # BIRBORB96803-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human TNFSF12 ELISA kit
Catalog # BIRBORB96803-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Human
  • Target protein:
    TNFSF12
  • Description:
    Human TNFSF12 ELISA Kit
  • Specificity:
    Natural and recombinant human TNFSF12
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human TNFSF12 ELISA Kit for the quantitative detection for human tumour necrosis factor ligand superfamily member 12 (TNFSF12).

  • No detectable cross-reactivity with other relevant proteins

Tumour necrosis factor ligand superfamily member 12 also known as TNF-related weak inducer of apoptosis TWEAK is a protein that in humans is encoded by the TNFSF12 gene. The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor TNF ligand family. This protein is a ligand for the FN14 or TWEAKR receptor. This cytokine has overlapping signaling functions with TNF, but displays a much wider tissue distribution. This cytokine can induce apoptosis via multiple pathways of cell death in a cell type-specific manner. This cytokine is also found to promote proliferation and migration of endothelial cells, and thus acts as a regulator ofangiogenesis. The TNFSF12 gene lies 878 bp upstream of the putative transcriptional start site of the TNFSF13 gene on chromosome 17p13.1.