HPLC columns, Polaris™ C8-ether
Supplier: VARIAN
Certificates
About this item
These Polaris™ phases have rapid high resolution separations based on ultra pure silica to which alkyl chains and polar modifications are attached.
- Enhanced retention and peak shape of polar analytes
- Additional selectivity for H-bond donors
- High aqueous compatibility
- No phase collapse
Frequently Bought Together