Pierce™ ABC Staining Kits
About this item
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
- Very low background staining
- Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.
4 Options Available Below
Product Details & Documents
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
- Very low background staining
- Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.
Documents
Product Family Options
Product Information
- PkAvailabilityPrice
- ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution1 KITLog in for availability1 * 1 KIT201 000 Ft
- PIER32050Ultra-sensitive ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solutionUltra-sensitive ABC peroxidase standard staining kit, sufficient for at least 10 ml of ABC staining solution1 KITLog in for availability1 * 1 KIT265 000 Ft
- PIER32052Ultra-sensitive ABC peroxidase mouse IgG staining kit, sufficient for at least 10 ml of ABC staining solutionUltra-sensitive ABC peroxidase mouse IgG staining kit, sufficient for at least 10 ml of ABC staining solution1 KITLog in for availability1 * 1 KIT380 000 Ft
- PIER32054Ultra-sensitive ABC peroxidase rabbit IgG staining kit, sufficient for at least 10 ml of ABC staining solutionUltra-sensitive ABC peroxidase rabbit IgG staining kit, sufficient for at least 10 ml of ABC staining solution1 KITLog in for availability1 * 1 KIT379 000 Ft