Order Entry
Hungary
ContactUsLinkComponent
Sodium acetate trihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Sodium acetate trihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Sodium acetate trihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • ACS - Assay (CH₃COONa·3H₂O):
    99.0 - 101.0 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤0.002 %
  • ACS - pH of 5% Solution at 25°C:
    7.5 - 9.2

Specification Test Results

Related Information

Frequently Bought Together