Anti-IL2RB Rabbit Polyclonal Antibody
BSBTPB9761
- Antibody type:Primary
- Antigen symbol:IL2RB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:16185
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Interleukin-2 receptor subunit beta(IL2RB) detection. Tested with WB in Mouse.
Type: Primary
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse