Anti-HFM1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-15471R
Supplier: Bioss
Specifications
- Antitest típus:Elsődleges
- Antigén szimbólum:HFM1
- Clonality:Polyclonal
- Konjugáció:Unconjugated
- Házigazda:Rabbit
- ImmunKémia:Yes
- Izotípus:IgG
- Reactivity:Human
- Western blot:Yes
- Forma:liquid
- Antigén szinonimák:HFM1|HFM1_HUMAN|Probable ATP-dependent DNA helicase HFM1|SEC63 domain-containing protein 1.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Koncentráció:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Méret:100 μg
- Kiszerelés:100 µl
Specifications
About this item
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HFM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human