Order Entry
Hungary
ContactUsLinkComponent
Peqlab, Components for Peqlab DNA Polymerases
Peqlab, Components for Peqlab DNA Polymerases
Catalog # PEQL01-1018
Supplier:  VWR Peqlab
CAS Number:  
Peqlab, Components for Peqlab DNA Polymerases
Catalog # PEQL01-1018
Supplier:  VWR Peqlab
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Application:
  • Description:
    10X PCR Buffer, incomplete
  • Environmentally Preferable:

Specifications

About this item