Order Entry
Hungary
ContactUsLinkComponent
VWR® Gold-N-Gel™, RNA dye
VWR® Gold-N-Gel™, RNA dye
Catalog # N734-5ML
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
VWR® Gold-N-Gel™, RNA dye
Catalog # N734-5ML
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    RNA dye (200X)
  • Seq:
    01
  • Environmentally Preferable:
    Safer Human and Environmental Health
Specifications
About this item
Gold-N-Gel RNA dye is compatible with standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illumination with UV light. A standard ethidium bromide filter can be used for gel documentation.
  • Non mutagenic, fluorescent in-gel stain for immediate visualisation of RNA bands on denaturing agarose gels
  • Immediate band visualisation with UV illumination - No post-run staining or destaining
  • Easy-to-use: Simply add to melted agarose prior to casting
Delivery: Supplied as a 200X concentrate.