Anti-CORIN Mouse Monoclonal Antibody [clone: 5B6]
USBI125235
- Antibody type:Primary
- Antigen name:Corin, serine peptidase
- Antigen symbol:CORIN
- Clonality:Monoclonal
- Clone:5B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:616 to 716
- Storage buffer:PBS, pH 7,2.
- Sequence:CKERDLWECPSNKQCLKHTVICDGFPDCPDYMDEKNCSFCQDDELECANHACVSRDLWCDGEADCSDSSDEWDCVTLSINVNSSSFLMVHRAATEHHVCA
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CORIN (corin, serine peptidase)
Clonality: monoclonal
Clone: 5B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human