Order Entry
Hungary
ContactUsLinkComponent
Anti-STAR Rabbit Polyclonal Antibody (Biotin)
Anti-STAR Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI042371-BIOTIN
Supplier:  US Biological
Anti-STAR Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI042371-BIOTIN
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Steroidogenic Acute Regulatory Protein
  • Antigen symbol:
    STAR
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    87 to 117
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: STAR (steroidogenic acute regulatory protein)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human