Anti-EPB41L4A Goat polyclonal antibody unconjugated
EVBIEB08238
- Antibody type:Primary
- Antigen name:Erythrocyte membrane protein band 4.1 like 4A
- Antigen symbol:EPB41L4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: EPB41L4A (erythrocyte membrane protein band 4.1 like 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse