Anti-XBP1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The XBP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to XBP1. This antibody reacts with human, mouse. The XBP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: XBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-77681SS
- NOVUNBP1-77681
- Antigen symbol:
- XBP1
- XBP1
- Antigen name:
- X-box Binding Protein 1
- X-box Binding Protein 1
- Antigen synonyms:
- XBP-1|TREB-5|TREB5|XBP2
- XBP-1|TREB-5|TREB5|XBP2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7494
- 7494
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together