Anti-LDL Receptor Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-06709V
- Antibody type:Primary
- Antigen name:low density lipoprotein receptor
- Antigen symbol:LDLR
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:3949
- Antigen synonyms:FHC|FH|LDLCQ2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: LDL Receptor
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse