About this item
The Dopamine D1 R / DRD1 Antibody (SG2-D1a) from Novus Biologicals is a mouse monoclonal antibody to Dopamine D1 R / DRD1. This antibody reacts with mouse, rat, bovine (negative), canine (negative), human (negative), porcine (negative), rabbit (negative), sheep (negative). The Dopamine D1 R / DRD1 Antibody (SG2-D1a) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: DRD1
Clonality: Monoclonal
Clone: SG2-D1a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative)
Specifications
- Catalog No:
- NOVUNB110-60017
- NOVUNB110-60017SS
- Antigen symbol:
- DRD1
- DRD1
- Antigen name:
- Dopamine receptor D1
- Dopamine receptor D1
- Antigen synonyms:
- DRD1A|DADR
- DRD1A|DADR
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- SG2-D1a
- SG2-D1a
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 1812
- 1812
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice