Order Entry
Hungary
ContactUsLinkComponent
Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
Catalog # ABCAAB138364-100
Supplier:  Abcam
undefined
Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
Catalog # ABCAAB138364-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PI3 Kinase p85 beta
  • Antigen symbol:
    p85 beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal to PI 3 Kinase p85 beta (phospho Y464).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase p85 beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human