Order Entry
Hungary
ContactUsLinkComponent
Anti-CSNK2A1 (phospho S362 + T360) Rabbit Polyclonal Antibody
Anti-CSNK2A1 (phospho S362 + T360) Rabbit Polyclonal Antibody
Catalog # ABCAAB119410-100
Gyártó:  Abcam
undefined
Anti-CSNK2A1 (phospho S362 + T360) Rabbit Polyclonal Antibody
Catalog # ABCAAB119410-100
Gyártó:  Abcam

Specifikációk

  • Antibody type:
    Primary
  • Antigen name:
    Casein Kinase 2 Alpha
  • Antigen symbol:
    CSNK2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifikációk

Termék információk

Rabbit polyclonal to CSNK2A1 (phospho T360 + S362).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CSNK2A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: